Anti-HMGN1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
USBI036644-APCEA 852 EUR
USBI036644-APC
Anti-HMGN1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: HMGN1 (high mobility group nucleosome binding domain 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR