Anti-HMGN5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
USBI036648-APCEA 852 EUR
USBI036648-APC
Anti-HMGN5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: HMGN5 (high mobility group nucleosome binding domain 5)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human