Anti-KCNJ15 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
USBI037326-BIOTINEA 852 EUR
USBI037326-BIOTIN
Anti-KCNJ15 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: KCNJ15 (potassium inwardly-rectifying channel, subfamily J, member 15)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human