Anti-KCNJ18 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
USBI037327-PEEA 852 EUR
USBI037327-PE
Anti-KCNJ18 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: KCNJ18 (potassium inwardly-rectifying channel, subfamily J, member 18)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human