Anti-KCNJ8 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: US Biological
USBI037331-HRPEA 852 EUR
USBI037331-HRP
Anti-KCNJ8 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: KCNJ8 (potassium inwardly-rectifying channel, subfamily J, member 8)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR