Anti-MATN1 Rabbit Polyclonal Antibody

Lieferant: US Biological
038222
USBI038222EA 587 EUR
USBI038222
Anti-MATN1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: MATN1 (matrilin 1, cartilage matrix protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR