Anti-MBD3L3 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
USBI038237-BIOTINEA 852 EUR
USBI038237-BIOTIN
Anti-MBD3L3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50
Type: Primary
Antigen: MBD3L3 (methyl-CpG binding domain protein 3-like 3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human