Anti-MCAM Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
038260-PE
USBI038260-PEEA 852 EUR
USBI038260-PE
Anti-MCAM Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Do not freeze!

Type: Primary
Antigen: MCAM (melanoma cell adhesion molecule)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR