Anti-MDK Rabbit Polyclonal Antibody

Lieferant: US Biological
038301
USBI038301EA 587 EUR
USBI038301
Anti-MDK Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: MDK (midkine (neurite growth-promoting factor 2))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR