Anti-MAP2K4 Rabbit Polyclonal Antibody

Lieferant: US Biological
038344
USBI038344EA 587 EUR
USBI038344
Anti-MAP2K4 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100

Type: Primary
Antigen: MAP2K4 (mitogen-activated protein kinase kinase 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR