Anti-MESP1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
USBI038364-APCEA 852 EUR
USBI038364-APC
Anti-MESP1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MESP1 (mesoderm posterior 1 homolog (mouse))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse