Anti-MET Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
038367-BIOTIN
USBI038367-BIOTINEA 852 EUR
USBI038367-BIOTIN
Anti-MET Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: MET (met proto-oncogene)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR