Anti-MEX3C Rabbit Polyclonal Antibody

Lieferant: US Biological
038393
USBI038393EA 587 EUR
USBI038393
Anti-MEX3C Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: MEX3C (mex-3 RNA binding family member C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR