Anti-MID1IP1 Rabbit Polyclonal Antibody

Lieferant: US Biological
USBI038428EA 587 EUR
USBI038428
Anti-MID1IP1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: MID1IP1 (MID1 interacting protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR