Anti-KMT2E Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
USBI038478-PEEA 852 EUR
USBI038478-PE
Anti-KMT2E Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Do not freeze!
Type: Primary
Antigen: KMT2E (lysine (K)-specific methyltransferase 2E)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human