Anti-MN1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
038518-BIOTIN
USBI038518-BIOTINEA 852 EUR
USBI038518-BIOTIN
Anti-MN1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: MN1 (meningioma (disrupted in balanced translocation) 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR