Anti-MOGT1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
USBI038533-PEEA 852 EUR
USBI038533-PE
Anti-MOGT1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: MOGT1
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human