Anti-MPP3 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
038565-BIOTIN
USBI038565-BIOTINEA 852 EUR
USBI038565-BIOTIN
Anti-MPP3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: MPP3 (membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR