Anti-MR1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
038575-PE
USBI038575-PEEA 852 EUR
USBI038575-PE
Anti-MR1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Do not freeze!

Type: Primary
Antigen: MR1 (major histocompatibility complex, class I-related)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR