Anti-MRPL28 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
USBI038593-BIOTINEA 852 EUR
USBI038593-BIOTIN
Anti-MRPL28 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MRPL28 (mitochondrial ribosomal protein L28)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human