Anti-MTCH2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: US Biological
USBI038651-APEA 852 EUR
USBI038651-AP
Anti-MTCH2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: MTCH2 (mitochondrial carrier 2)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse