Anti-MTMR4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
USBI038668-APCEA 852 EUR
USBI038668-APC
Anti-MTMR4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MTMR4 (myotubularin related protein 4)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human