Anti-MTMR4 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
USBI038668-BIOTINEA 852 EUR
USBI038668-BIOTIN
Anti-MTMR4 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MTMR4 (myotubularin related protein 4)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human