Anti-MTMR6 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
USBI038669-PEEA 852 EUR
USBI038669-PE
Anti-MTMR6 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MTMR6 (myotubularin related protein 6)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human