Anti-MTNR1A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
USBI038672-APCEA 678 EUR
USBI038672-APC
Anti-MTNR1A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: MTNR1A (melatonin receptor 1A)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human