Anti-MTNR1A Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
USBI038672-PEEA 678 EUR
USBI038672-PE
Anti-MTNR1A Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: MTNR1A (melatonin receptor 1A)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human