Anti-MUC1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
038690-BIOTIN
USBI038690-BIOTINEA 852 EUR
USBI038690-BIOTIN
Anti-MUC1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Dot blot 1:500; Immunofluorescence: 1:10-50

Type: Primary
Antigen: MUC1 (mucin 1, cell surface associated)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR