Anti-MUC20 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: US Biological
USBI038694-FITCEA 852 EUR
USBI038694-FITC
Anti-MUC20 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50
Do not freeze!
Type: Primary
Antigen: MUC20 (mucin 20, cell surface associated)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human