Anti-MURC Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
USBI038701-BIOTINEA 852 EUR
USBI038701-BIOTIN
Anti-MURC Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Type: Primary
Antigen: MURC (muscle-related coiled-coil protein)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse