Anti-MYC Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
USBI038719-APEA 852 EUR
USBI038719-AP
Anti-MYC Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA1:1000 Dot blot 1:500
Do not freeze!

Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR