Anti-MPO Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: US Biological
038726-HRP
USBI038726-HRPEA 852 EUR
USBI038726-HRP
Anti-MPO Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR