Anti-MYF5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: US Biological
USBI038728-HRPEA 852 EUR
USBI038728-HRP
Anti-MYF5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MYF5 (myogenic factor 5)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human