Anti-MYLIP Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
USBI038739-PEEA 852 EUR
USBI038739-PE
Anti-MYLIP Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: MYLIP (myosin regulatory light chain interacting protein)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR