Anti-MYO1E Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
USBI038745-PEEA 852 EUR
USBI038745-PE
Anti-MYO1E Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: MYO1E (myosin IE)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR