Anti-NAAA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: US Biological
038775-HRP
USBI038775-HRPEA 852 EUR
USBI038775-HRP
Anti-NAAA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NAAA (N-acylethanolamine acid amidase)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR