Anti-NAP1L2 Rabbit Polyclonal Antibody

Lieferant: US Biological
038790
USBI038790EA 587 EUR
USBI038790
Anti-NAP1L2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NAP1L2 (nucleosome assembly protein 1-like 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR