Anti-NARFL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: US Biological
USBI038796-FITCEA 852 EUR
USBI038796-FITC
Anti-NARFL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: NARFL (nuclear prelamin A recognition factor-like)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR