Anti-NAA15 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
038797-PE
USBI038797-PEEA 852 EUR
USBI038797-PE
Anti-NAA15 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: NAA15 (N(alpha)-acetyltransferase 15, NatA auxiliary subunit)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR