Anti-NAT8 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
038810-APC
USBI038810-APCEA 852 EUR
USBI038810-APC
Anti-NAT8 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: NAT8 (N-acetyltransferase 8 (GCN5-related, putative))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR