Anti-NDUFA12 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
USBI038857-BIOTINEA 852 EUR
USBI038857-BIOTIN
Anti-NDUFA12 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: NDUFA12 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human