Anti-NEK3 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
038884-BIOTIN
USBI038884-BIOTINEA 852 EUR
USBI038884-BIOTIN
Anti-NEK3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: NEK3 (NIMA-related kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR