Anti-NPHS1 Rabbit Polyclonal Antibody

Lieferant: US Biological
038897
USBI038897EA 587 EUR
USBI038897
Anti-NPHS1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Dot blot 1:100-500

Type: Primary
Antigen: NPHS1 (nephrosis 1, congenital, Finnish type (nephrin))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR