Anti-NEUROD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: US Biological
038911-HRP
USBI038911-HRPEA 852 EUR
USBI038911-HRP
Anti-NEUROD1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:100

Type: Primary
Antigen: NEUROD1 (neuronal differentiation 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR