Anti-NF kappa B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: US Biological
USBI038926-FITCEA 852 EUR
USBI038926-FITC
Anti-NF kappa B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Recommended dilution: FLISA: 1:1000; Dot blot 1:100-500
Do not freeze!
Type: Primary
Antigen: NF kappa B
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human