Anti-NF kappa B Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
038926-PE
USBI038926-PEEA 852 EUR
USBI038926-PE
Anti-NF kappa B Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Dot blot 1:100-500
Do not freeze!

Type: Primary
Antigen: NF kappa B
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR