Anti-NGRN Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
USBI038957-BIOTINEA 852 EUR
USBI038957-BIOTIN
Anti-NGRN Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: NGRN (neugrin, neurite outgrowth associated)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR