Anti-NIPSNAP3B Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
038978-AP
USBI038978-APEA 852 EUR
USBI038978-AP
Anti-NIPSNAP3B Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: NIPSNAP3B (nipsnap homolog 3B (C. elegans))
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR