Anti-NKAI4 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
USBI038982-BIOTINEA 852 EUR
USBI038982-BIOTIN
Anti-NKAI4 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NKAI4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR