Anti-NOC3L Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
039040-BIOTIN
USBI039040-BIOTINEA 852 EUR
USBI039040-BIOTIN
Anti-NOC3L Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NOC3L (nucleolar complex associated 3 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR