Anti-NPM3 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
USBI039093-BIOTINEA 852 EUR
USBI039093-BIOTIN
Anti-NPM3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: NPM3 (nucleophosmin/nucleoplasmin 3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR